Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite rod, 6.15mm (0.242 in.) dia. x 152mm (6 in.) long, 99.9995% (metals basis)

Chemical Identifiers

Chemical Name or Material Graphite rod
Name Note 6.15mm (0.242 in.) dia. x 152mm (6 in.) long
Purity Grade Notes SPK grade, ultra 'F' purity
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA40767KD

 
alfa aesar™
40767KD
50 pieces CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 50 pieces
Purity Grade Notes SPK grade, ultra 'F' purity
Chemical Name or Material Graphite rod
TSCA TSCA
Name Note 6.15mm (0.242 in.) dia. x 152mm (6 in.) long
Physical Form Rod
Assay 99.9995%
Assay Percent Notes (metals basis)
MDL Number MFCD00144065