Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite rod, 5.0mm (0.20 in.) dia., 99.997% (metals basis)

$390.97 - $390.97

Chemical Identifiers

Chemical Name or Material Graphite rod
Name Note 5.0mm (0.20 in.) dia.
Purity Grade UF-4S Grade
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45076DW

  SDS
alfa aesar™
45076DW
300mm Each for $390.97
Description & Specifications

Specifications

Quantity 300mm
Chemical Name or Material Graphite rod
TSCA TSCA
Name Note 5.0mm (0.20 in.) dia.
Purity Grade UF-4S Grade
Physical Form Rod
Assay 99.997%
Assay Percent Notes (metals basis)
MDL Number MFCD00144065