Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite rod, 10.0mm (0.40 in.) dia., 99.997% (metals basis)

Chemical Identifiers

Chemical Name or Material Graphite rod
Name Note 10.0mm (0.40 in.) dia.
Purity Grade UF-4S Grade
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA45077DW

 
alfa aesar™
45077DW
300mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 300mm
Chemical Name or Material Graphite rod
TSCA TSCA
Name Note 10.0mm (0.40 in.) dia.
Purity Grade UF-4S Grade
Physical Form Rod
Assay 99.997%
Assay Percent Notes (metals basis)
MDL Number MFCD00144065