missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

EISCO Graphite (Pure Carbon) Specimen

For use in classroom identification labs and exercises. EISCO Graphite (Pure Carbon) Specimen correlates well to Next Generation Science Standards Evidence Statements.

Manufacturer:  EISCO ESNG0009PK12

 View more versions of this product

Catalog No. S24670



Description

Description

  • Example of metallic luster
  • Representation of specimen that is pure carbon and not as expensive as diamond
  • Specimen comes in clear labeled and barcoded bag for ideal storage
  • Average specimen size: 2cm
  • Made of same material found in pencils
Specifications

Specifications

Clear labeled and barcoded bag
Study example of metallic luster
Approx. 1 in.
12/Pk.
12 specimen
Graphite (Pure Carbon) study specimen
Approx. 3cm
Documents