Learn More
Graphite powder, synthetic, APS 7-11 micron, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 04630414
Description
Graphite powder is basically used for casting polishing purposes and molding. Natural graphite is mostly used for refractories, batteries, steel making, expanded graphite, brake linings, foundry facings and lubricants. It is also used in pigments, anodes arc-lamp carbons, coating cathode ray tubes and in moderator in nuclear piles. Graphite (carbon) fiber and carbon nanotubes are also used in carbon fiber reinforced plastics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphite powder | |
7782-42-5 | |
C | |
25 g | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
99% |
Synthetic | |
Odorless | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
UN1325 | |
Insoluble in water as well as other organic solvents. | |
[C] | |
12.01 | |
CHEBI:27594 | |
Powder |
Chemical Identifiers
7782-42-5 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Safety and Handling
GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P304+P340-P305+P351+P338-P312-P370+P378q-P501c
H228-H319-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 231-95593
RTECSNumber : MD9659600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only