missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite powder, synthetic, APS 7-11 micron, 99%

Manufacturer:  Alfa Aesar™ 4630430

 View more versions of this product

Catalog No. AA4630430



Specifications

Specifications

Graphite powder
APS 7 to 11 Micron
Synthetic
C
UN1325
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Powder
Odorless
250g
7782-42-5
MFCD00144065
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.011
CHEBI:27594
99%

Chemical Identifiers

7782-42-5
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00144065
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : MD9659600

TSCA : TSCA

SDS Documents