Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite powder, natural, high purity, -200 mesh, 99.9999% (metals basis)

$93.42 - $93.42

Chemical Identifiers

Chemical Name or Material Graphite powder
Name Note Natural
Purity Grade Notes Ultra superior purity
MDL Number MFCD00144065
UN Number UN1325
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14734TA

  SDS
alfa aesar™
14734TA
28g Each for $93.42
Description & Specifications

Specifications

Quantity 28g
Purity Grade Notes Ultra superior purity
Chemical Name or Material Graphite powder
TSCA TSCA
Name Note Natural
Physical Form Powder
Assay 99.9999%
Assay Percent Notes (metals basis)
MDL Number MFCD00144065
UN Number UN1325
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
Mesh Size −200 Mesh
Odor Odorless
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.