missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite powder, natural, briquetting grade, -100 mesh, 99.9995% (metals basis)

Manufacturer:  Alfa Aesar™ 14735TB

 View more versions of this product

Catalog No. AA14735TB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Graphite powder
Odorless
113g
7782-42-5
UCP-1 grade, ultra 'F' purity
(metals basis)
MFCD00144065
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Powder
−100 Mesh
Black
Natural
Briquetting Grade
99.9995%
C
UN1325
[C]
12.011
CHEBI:27594

Chemical Identifiers

7782-42-5
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
CHEBI:27594
[C]
C
MFCD00144065
5462310
carbon
Safety and Handling

Safety and Handling

GHS H Statement
H228-H319-H335
Flammable solid.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.

RTECSNumber : MD9659600

TSCA : TSCA

SDS Documents