Please login to your online account to display your discounted pricing

Graphite powder, crystalline, -300 mesh, 99%, Alfa Aesar

Synthetic from desulfurized petroleum coke

$44.59 - $44.59

Chemical Identifiers

Chemical Name or Material Graphite
CAS 7782-42-5
Physical Form Crystalline Powder
Assay Percent Range 99%
Molecular Formula (CF)
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1012930

  SDS
alfa aesar
1012930
250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA10129A1

  SDS
alfa aesar
10129A1
1kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1012914

  SDS
alfa aesar
1012914
25g Each for $44.59
Description & Specifications

Specifications

Physical Form Crystalline Powder
Quantity 250g
Color Black
DOT Information DOT Class: 4.1, Packaging Group: III
EINECS Number 231-955-3
MDL Number MFCD00144065
Density ca 2g/cm3
CAS 7782-42-5
Odor Odorless
Assay Percent Range 99%
Appearance Black powder
Molecular Formula (CF)
Health Hazard 1 H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.
Chemical Name or Material Graphite
Safety and Handling

H228-H319-H335: Flammable solid. Causes serious eye irritation. May cause respiratory irritation.