Please login to your online account to display your discounted pricing

Graphite, powder, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC385031000

 View more versions of this product

Catalog No. AC385031000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 12.01
Quantity 100g
Color Black
Merck Index 15, 4575
Chemical Name or Material Graphite
Molecular Formula C
MDL Number MFCD00144065
CAS 7782-42-5
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Ash 0.8% max.
Carbon (C) 99.0% min.
Packaging Glass bottle
Sulfur (S) 0.05% max.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Graphite
CAS 7782-42-5
Molecular Formula C
Formula Weight 12.01
MDL Number MFCD00144065
Accessories

Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents