Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite plate, resin impregnated, 6.35mm (0.25 in.) thick

Chemical Identifiers

Chemical Name or Material Graphite plate
Name Note Resin impregnated, 6.35mm (0.25 in.) thick
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41831GH

 
alfa aesar™
41831GH
100x100mm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Graphite plate
TSCA TSCA
Name Note Resin impregnated, 6.35mm (0.25 in.) thick
MDL Number MFCD00144065
Quantity 100x100mm