missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Graphite Foil, 0.254mm (0.01 in.) thick, 99.8% (Metals basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 10832CG

 View more versions of this product

Catalog No. AA10832CG



Specifications

Specifications

Dark Gray to black foil
Gray to Black
7782-42-5
C
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.011
CHEBI:27594
Graphite
15 x 60cm
99.8% (Metals basis)
MFCD00144065
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Foil, 0.254mm (0.010 in.) thick

Chemical Identifiers

7782-42-5
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00144065
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]
Documents