missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Graphite Flake, Natural, -10 Mesh, 99.9% (Metal Basis), Alfa Aesar™

Manufacturer:  Alfa Aesar 4331918

 View more versions of this product

Catalog No. AA4331918



Specifications

Specifications

Graphite
50g
7782-42-5
C
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
[C]
12.011
CHEBI:27594
Undesignated
Natural
99.9%, metal basis
MFCD00144065
OKTJSMMVPCPJKN-UHFFFAOYSA-N
carbon
5462310
Flake, -10 Mesh

Chemical Identifiers

7782-42-5
12.011
OKTJSMMVPCPJKN-UHFFFAOYSA-N
5462310
carbon
C
MFCD00144065
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
CHEBI:27594
[C]
Documents