Please login to your online account to display your discounted pricing

Graphite felt, 99.9% (metal basis), Alfa Aesar™

Form: approx.9.2g/10 x 10cm

Chemical Identifiers

Chemical Name or Material Graphite felt
Physical Form Felt, 1.12cm (0.441 in.) thick
Assay Percent Range 99.9% (Metal basis)
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA14630RR

  SDS
alfa aesar
14630RR
10 x 10cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA14630RY

  SDS
alfa aesar
14630RY
25 x 25cm CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 10 x 10cm
Color Dark gray to black
Chemical Name or Material Graphite felt
EINECS Number 231-955-3
Physical Form Felt, 1.12cm (0.441 in.) thick
Assay Percent Range 99.9% (Metal basis)
MDL Number MFCD00144065
Appearance Dark grey to black solid in various forms