Please login to your online account to display your discounted pricing

Alfa Aesar™ Graphite adhesive, aqueous based

$51.49 - $51.49

Chemical Identifiers

Chemical Name or Material Graphite adhesive, aqueous based
Name Note Service temp: 306°C
MDL Number MFCD00144065
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4121418

 
alfa aesar™
4121418
50g Each for $51.49
Description & Specifications

Specifications

Quantity 50g
Chemical Name or Material Graphite adhesive, aqueous based
TSCA TSCA
Name Note Service temp: 306°C
Physical Form Flakes
MDL Number MFCD00144065
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Odor Ammonia-like
Particle Size 1 Micron
Color Black
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.