missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Graphene nanoplatelets aggregates, sub-micronparticles, S.A. 500 m^2/g, Thermo Scientific Chemicals
$63.79 - $192.56
Chemical Identifiers
CAS | 7782-42-5 |
---|---|
Molecular Formula | C |
Molecular Weight (g/mol) | 12.01 |
MDL Number | MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 |
InChI Key | OKTJSMMVPCPJKN-UHFFFAOYSA-N |
Synonym | graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve |
PubChem CID | 5462310 |
ChEBI | CHEBI:27594 |
IUPAC Name | carbon |
SMILES | [C] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7782-42-5 | |
12.01 | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
5462310 | |
carbon |
C | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
CHEBI:27594 | |
[C] |
Specifications
7782-42-5 | |
C | |
25 g | |
OKTJSMMVPCPJKN-UHFFFAOYSA-N | |
carbon | |
5462310 | |
Nanoplatelets |
Black | |
MFCD00133992 MFCD00144065 MFCD00146977 MFCD00211867 MFCD06411993 MFCD07370731 | |
graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve | |
[C] | |
12.01 | |
CHEBI:27594 | |
Graphene nanoplatelets aggregates, sub-micronparticles |
Safety and Handling
GHS H Statement
H319-H335
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
EINECSNumber : 231-955-3
RTECSNumber : MD9659600
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only