missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Graphene, nanoplatelets 2-10 nm, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 453160500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Graphene | |
3650.0°C | |
MFCD00144065 | |
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon | |
VNWKTOKETHGBQD-UHFFFAOYSA-N | |
methane | |
297 | |
12.01 |
1034343-98-0 | |
Cx | |
50 g | |
Solubility in water: insoluble. | |
C | |
16.04 | |
CHEBI:16183 |
Chemical Identifiers
1034343-98-0 | |
16.04 | |
VNWKTOKETHGBQD-UHFFFAOYSA-N | |
297 | |
methane |
Cx | |
MFCD00144065 | |
methyl hydride, marsh gas, biogas, fire damp, r 50 refrigerant, methan, metano, hsdb 167, in gaseus state, tetrahydridocarbon | |
CHEBI:16183 | |
C |
RUO – Research Use Only