missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Granzyme M Mouse anti-Human, eFluor 660, Clone: 4B2G4, eBioscience™

Mouse Monoclonal Antibody

$216.05 - $520.55

Specifications

Antigen Granzyme M
Clone 4B2G4
Host Species Murine
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501124541 Life Technologies
LS50977441
25 Tests Each for $216.05
501124542 Life Technologies
LS50977442
100 Tests Each for $520.55
Specifications

Description

Granzyme M is one of five granzyme serine proteases that have been indentified in humans. These proteins are expressed in the granules of NK cells and cytotoxic T cells, and are critical for the induction of target cell apoptosis through the cleavage of intracellular substrates. Although not as well characterized as some of the other family members, Granzyme M has been found to exhibit some distinct functions. These include the non-cytotoxic inhibition of cytomegalovirus replication, induction of apoptosis in tumor cells, and contribution to the TLR4-induced inflammatory process.
Description & Specifications

Specifications

Granzyme M
Murine
Flow Cytometry (Intracellular Staining)
eFluor 660
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test
4B2G4
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer