Please login to your online account to display your discounted pricing

Granzyme K Mouse anti-Human, PerCP-eFluor 710, Clone: G3H69, eBioscience™

Mouse Monoclonal Antibody

$172.55 - $419.05

Specifications

Antigen Granzyme K
Clone G3H69
Host Species Mouse
Gene Alias GZMK, Granzyme 3, TRYP2
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123921

 
life technologies
LS46889741
25 tests Each for $172.55

501123922

 
life technologies
LS46889742
100 tests Each for $419.05
Description & Specifications

Specifications

Antigen Granzyme K
Clone G3H69
Host Species Mouse
Gene Alias GZMK, Granzyme 3, TRYP2
Species Reactivity Human
Applications Flow Cytometry (Intracellular Staining)
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 5μL (0.03μg)/test

This G3H69 monoclonal antibody reacts with human Granzyme K. Granzyme K is one of five granzyme serine proteases that have been identified in humans. These proteins are expressed in the granules of NK cells and cytotoxic T cells, and are critical for the induction of target cell apoptosis through the cleavage of intracellular substrates. Granzyme A and Granzyme K are both tryptases and appear to show overlapping, though not identical, substrate specificity. This functional similarity is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTLs.