missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Granzyme B, clone: 16G6, eBioscience™

Rat Monoclonal Antibody

Manufacturer:  Life Technologies LS14882282

 View more versions of this product

Catalog No. 501129084



Description

Description

Granzyme B is a member of the granzyme serine protease family. Granzyme B is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation, and CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. Granzyme B is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for Granzyme B during cytotoxic T cell-induced apoptosis.

Description: The 16G6 antibody reacts with mouse Granzyme B (GrB) which is a member of the granzyme serine protease family. GrB is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation. GrB has been called CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. GrB is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for granzyme B during cytotoxic T cell-induced apoptosis.For intracellular staining and flow cytometric analysis with direct conjugates of anti-mouse Granzyme B, it is highly recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523). Other buffers may yield varying results. For more information, please contact technical support at tech@ebioscience.com.Applications Reported: This 16G6 antibody has been reported for use in western blotting, immunohistochemical staining, ELISA capture antibody, and immunohistochemical staining of formalin-fixed paraffin embedded tissue sections.Applications Tested: The unlabelled 16G6 antibody has been tested as the capture antibody in a sandwich ELISA for quantitation of mouse granzyme B protein levels, in combination with the biotinylated eBioLUEE (13-8821) antibody for detection and recombinant mouse granzyme B as the standard. A suitable range of concentrations of this antibody for ELISA detection is 1-4 μg/mL. The 16G6 antibody has been tested for Western blotting; a recommended starting concentration is 1 μg/mL. The 16G6 antibody has been used for formalin-fixed paraffin embedded tissue using low pH antigen retrieval at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest.Purity: Greater than 90%, as determined by SDS-PAGE.Aggregation: Less than 10%, as determined by HPLC.Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

Granzyme B
Monoclonal
0.5mg/mL
PBS with 0.09% sodium azide; pH 7.2
P04187
GrB
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Murine
ELISA, Immunohistochemistry (Paraffin), Western Blot, Western Blot
16G6
Unconjugated
Gzmb
Liquid
Gzmb
IgG2b, kappa, kappa
100μg
Store at 2-8°C.
Primary
14939
Documents

For Research Use Only.