Please login to your online account to display your discounted pricing

Granzyme B Rat anti-Mouse, Biotin, Clone: 16G6, eBioscience™

Rat Monoclonal Antibody

$158.05 - $404.55

Specifications

Antigen Granzyme B
Clone 16G6
Host Species Rat
Gene Alias GrzB, GrB
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5012219

 
life technologies
LS13882280
25μg Each for $158.05

501129153

 
life technologies
LS13882282
100μg Each for $404.55
Description & Specifications

Specifications

Antigen Granzyme B
Clone 16G6
Host Species Rat
Gene Alias GrzB, GrB
Species Reactivity Mouse
Applications Flow Cytometry (Intracellular Staining)
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blotting
Regulatory Status RUO
Conjugate Biotin
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

The 16G6 antibody reacts with mouse Granzyme B (GrB) which is a member of the granzyme serine protease family. GrB is found in the granules of cytotoxic T cells and NK cells. Granzyme B has also been described as CGL1 (cathepsin G-like-1), a serine protease expressed only in cytotoxic T-lymphocytes after cell activation. GrB has been called CTLA-1 (cytotoxic T lymphocyte-associated serine esterase 1) based on identification of mRNA in various cytotoxic T cells, but not observed in non-cytotoxic lymphoid cells. GrB is crucial for the rapid induction of target cell death by apoptosis, induced by interaction with cytotoxic T cells. The receptor involved has been identified as mannose 6-phosphate receptor. This receptor functions as a death receptor for granzyme B during cytotoxic T cell-induced apoptosis.

For intracellular staining and flow cytometric analysis with direct conjugates of anti-mouse Granzyme B, it is highly recommended to use the Foxp3 Staining Buffer Set (cat. 00-5523). Other buffers may yield varying results. For more information, please contact technical support at tech@ebioscience.com.