missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Granzyme A Mouse anti-Human, PE, Clone: CB9, eBioscience™

Mouse Monoclonal Antibody

$157.08 - $363.00

Specifications

Antigen Granzyme A
Clone CB9
Host Species Murine
Gene Alias CTLA-3, Fragmentin-1, TSP-1
Species Reactivity Human
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501122426 Life Technologies
LS12917741
25 Tests Each for $157.08
501122427 Life Technologies
LS12917742
100 Tests Each for $363.00
Specifications

Description

This CB9 monoclonal antibody reacts with human Granzyme A. Granzyme A is the most abundantly expressed of the granzyme serine proteases, which are proteins released from the granules of NK cells and cytotoxic T lymphocytes that induce death in target cells by cleavage of intracellular substrates. They play a critical role in immune defense against viruses, tumors, and intracellular bacteria. Granzyme A activates caspase-independent cell death pathways morphologically similar to apoptosis and characterized by mitochondrial and DNA damage. It may also play a role in inflammation, as the precursor form of IL-1 beta (pro-IL-1 beta) is among its target substrates. Granzyme A shares overlapping substrate specificity with the closely-related Granzyme K, which is believed to account for the minimal decrease in cytotoxicity of Granzyme A-deficient CTL.
Description & Specifications

Specifications

Granzyme A
Murine
Human
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
CB9
CTLA-3, Fragmentin-1, TSP-1
Flow Cytometry (Intracellular Staining)
PE
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
5μL (0.125μg)/test