Please login to your online account to display your discounted pricing

Gramine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Gramine
CAS 87-52-5
Assay ≥98.5% (on anhydrous substance)
Assay Percent Range 99%
Molecular Formula C11H14N2
Formula Weight 174.25
MDL Number MFCD00005629
Synonym 3-(Dimethylaminomethyl)indole
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC120180100

  SDS
thermo fisher scientific
120180100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC120181000

  SDS
thermo fisher scientific
120181000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC120185000

  SDS
thermo fisher scientific
AC120185000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.5% (on anhydrous substance)
Formula Weight 174.25
Physical Form Powder
Quantity 10g
Color Beige
Merck Index 15, 4569
Infrared Spectrum Authentic
Synonym 3-(Dimethylaminomethyl)indole
Sulfated Ash 0.5% max.
Chemical Name or Material Gramine
Loss on Drying 0.5% max.
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C11H14N2
MDL Number MFCD00005629
CAS 87-52-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.