Please login to your online account to display your discounted pricing

Gram's Iodine, Dilute Lugol's Iodine, for Gram Staining, Ricca Chemical


Quantity 100mL
Packaging Amber Glass Bottle

Manufacturer: ricca chemical company  RIR3450000100C

 View more versions of this product

Catalog No. R3450000100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Color Dark brown
Quantity 100mL
CAS Min % 0.36
Chemical Name or Material Gram's Iodine
CAS 7732-18-5
Name Note Dilute Lugol's Iodine
CAS Min % 0.73
Packaging Amber Glass Bottle
CAS 7681-11-0
CAS Max % 0.76
CAS Max % 100.0
CAS 7553-56-2
CAS Max % 0.38
CAS Min % 97.0

Specifications

Chemical Name or Material Gram's Iodine
Name Note Dilute Lugol's Iodine
CAS 7732-18-5
CAS Min % 97.0
CAS Max % 100.0
CAS 7681-11-0
CAS Min % 0.73
CAS Max % 0.76
CAS 7553-56-2
CAS Min % 0.36
CAS Max % 0.38
Physical Form Liquid