Please login to your online account to display your discounted pricing
Graham Field Stainless-Steel Blades for Dissecting Knife Handles
Exceptionally strong and uniformly sharp
Manufacturer: feather safety razor co  NO.12
 View more versions of this product
Catalog No. 08-916-5C
  • / Pack of 100 Each
Description & Specifications
Specifications
Size 12
Length (English) 1.37 in.
For Use With (Equipment) For handle sizes 3, 5, 7
Length (Metric) 3.5cm
Sterile Yes
Type Blade
Material Stainless Steel
Accessories
Accessories for Catalog Number: 089165C
Item Mfr. No. Item Description Price Per Unit Quantity