GSC Intenational Graduated Resonance Tube
Choose GSC tubes for demonstrating resonance in the classroom.
Manufacturer:  GSC Intenational GIG18032
 View more versions of this product
Catalog No. S42091
Specifications
Description
Resonance Tube; GSC; Inexpensive way to demonstrate resonance; Open-ended glass tubes are annealed and fire-polished with silk- screened centimeter scale; 4 x 60cm w/scale
An inexpensive way to demonstrate resonance. These open-ended glass tubes are annealed and fire-polished with silk- screened centimeter scale.
Description & Specifications
Specifications
60cm
Resonance Tube
23.6 in.
Metric graduated scale