Please login to your online account to display your discounted pricing

Qorpak™ Graduated Medium Round Bottle Beakers™ without Caps

For mixing, measuring and storage

$65.71 - $796.46

Specifications

Closure Screw
Color Clear
Material Glass
View More Specs
Products
Catalog Number Mfr. No. Capacity Height (English) Quantity Height (Metric) Price Quantity    

02991620

 
qorpak™
GLA00840
1 oz., 30mL 2.68 in. 432/Cs. 68mm Case for $796.46

02991715

 
qorpak™
GLA00846
8 oz., 240mL 5 in. 96/Cs. 127mm Case for $291.49

02991732

 
qorpak™
GLA00848
16 oz., 480mL 5.71 in. 48/Cs. 145mm Case for $191.60

02991796

 
qorpak™
GLA00844
4 oz., 120mL 4.02 in. 144/Cs. 102mm Case for $327.94

02991815

 
qorpak™
GLA00842
2 oz., 60mL 3.27 in. 288/Cs. 83mm Case for $615.34

02991952

 
qorpak™
GLA00841
1 oz., 30mL 2.68 in. 48/Cs. 68mm Case for $101.46

02992152

 
qorpak™
GLA00849
16 oz., 480mL 5.71 in. 24/Cs. 145mm Case for $117.31

02992800

 
qorpak™
GLA00845
4 oz., 120mL 4.02 in. 24/Cs. 102mm Case for $65.71

02992749

 
qorpak™
GLA00843
2 oz., 60mL 3.27 in. 48/Cs. 83mm Case for $109.29

02992920

 
qorpak™
GLA00847
8 oz., 240mL 5 in. 24/Cs. 127mm Case for $84.66
Description & Specifications

Specifications

Closure Screw
Color Clear
Material Glass

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents