Please login to your online account to display your discounted pricing

American Educational Products Graduated Measuring Jugs


Graduated measuring and sturdy handles.

Manufacturer: american educational products  700-500

 View more versions of this product

Catalog No. S95160

  • / Pack of 12 Each
 

Description & Specifications

Specifications

Capacity 16.9 oz., 500mL
Capacity (Metric) 500mL
Capacity (English) 16.9 oz.
Color Translucent
Includes Sturdy handle, molded raised graduation
Material Polypropylene

Translucent polypropylene measuring jugs incorporate a reinforced, sturdy handle.

  • Molded, raised graduations
  • Chemically resistant
  • Autoclavable