Please login to your online account to display your discounted pricing
American Educational Products Graduated Measuring Jugs
Graduated measuring and sturdy handles.
$44.95 - $44.95
Specifications
Color Translucent
Material Polypropylene
Includes Sturdy handle, molded raised graduation
View More Specs
Products
Catalog Number Mfr. No. Capacity Price Quantity    
S95160
american educational products
700-500
16.9 oz., 500mL Pack of 12 Each for $44.95
Description & Specifications
Specifications
Color Translucent
Material Polypropylene
Includes Sturdy handle, molded raised graduation
Translucent polypropylene measuring jugs incorporate a reinforced, sturdy handle.
  • Molded, raised graduations
  • Chemically resistant
  • Autoclavable