Please login to your online account to display your discounted pricing

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Solid PE Caps

For mixing, measuring and storage

$101.14 - $1,133.46

Specifications

Closure Liner Polyethylene
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Type Quantity Closure Size Price Quantity    

02991875

 
qorpak™
GLC05024
1 oz., 30mL Bulk Pack 432/Cs. 33-400 Case for $1,133.46

02991980

 
qorpak™
GLC01545
8 oz., 240mL Bulk Pack 96/Cs. 58-400 Case for $468.46

02992099

 
qorpak™
GLC01516
4 oz., 120mL Bulk Pack 144/Cs. 48-400 Case for $520.63

02992111

 
qorpak™
GLC01569
16 oz., 480mL Bulk Pack 48/Cs. 70-400 Case for $321.09

02992124

 
qorpak™
GLC01486
2 oz., 60mL Bulk Pack 288/Cs. 38-400 Case for $904.37

02992134

 
qorpak™
GLC01488
2 oz., 60mL Conventional Pack 48/Cs. 38-400 Case for $177.29

02992139

 
qorpak™
GLC01462
1 oz., 30mL Conventional Pack 48/Cs. 33-400 Case for $142.71

02992691

 
qorpak™
GLC01518
4 oz., 120mL Conventional Pack 24/Cs. 48-400 Case for $101.14

02992810

 
qorpak™
GLC01547
8 oz., 240mL Conventional Pack 24/Cs. 58-400 Case for $127.74

02992928

 
qorpak™
GLC01571
16 oz., 480mL Conventional Pack 24/Cs. 70-400 Case for $167.54
Description & Specifications

Specifications

Closure Liner Polyethylene
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Clear
Material Glass

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents