missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Solid PE Caps
For mixing, measuring and storage
Supplier: Qorpak™ GLC01488
Description
- Designed with slight shoulder for easy pouring and handling
- Molded graduations in mL and ounces
- Made of clear glass
- For use with biological and pathological specimens
- For storage of industrial laboratory chemicals and reagents
Specifications
Phenolic | |
42 mm | |
Screw Cap | |
38-400 mm | |
60 mL | |
Clear | |
Glass | |
48/Cs. |
83 mm | |
3.27 in. | |
Polyethylene | |
2 oz., 60 mL | |
2 oz. | |
1.65 in. | |
Conventional Pack | |
Black |