Please login to your online account to display your discounted pricing

Qorpak™ Graduated Glass Bottle Beakers with Black Phenolic Pulp/Tin Foil Caps

For mixing, measuring and storage

$122.77 - $1,089.23

Specifications

Closure Liner Pulp/Tin Foil
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Clear
View More Specs
Products
Catalog Number Mfr. No. Capacity Packaging Type Quantity Closure Size Price Quantity    

02991958

 
qorpak™
GLC01489
2 oz., 60mL Bulk Pack 288/Cs. 38-400 Case for $1,089.23

02992137

 
qorpak™
GLC01463
1 oz., 30mL Conventional Pack 48/Cs. 33-400 Case for $172.26

02992225

 
qorpak™
GLC01548
8 oz., 240mL Bulk Pack 96/Cs. 58-400 Case for $586.80

02992282

 
qorpak™
GLC01572
16 oz., 480mL Bulk Pack 48/Cs. 70-400 Case for $402.77

02992331

 
qorpak™
GLC01519
4 oz., 120mL Bulk Pack 144/Cs. 48-400 Case for $649.09

02992634

 
qorpak™
GLC01490
2 oz., 60mL Conventional Pack 48/Cs. 38-400 Case for $199.43

02992739

 
qorpak™
GLC01520
4 oz., 120mL Conventional Pack 24/Cs. 48-400 Case for $122.77

02992878

 
qorpak™
GLC01549
8 oz., 240mL Conventional Pack 24/Cs. 58-400 Case for $156.94

02992927

 
qorpak™
GLC01573
16 oz., 480mL Conventional Pack 24/Cs. 70-400 Case for $208.89
Description & Specifications

Specifications

Closure Liner Pulp/Tin Foil
Closure Material Phenolic
Closure Screw
Closure Color Black
Color Clear
Material Glass

  • Designed with slight shoulder for easy pouring and handling
  • Molded graduations in mL and ounces
  • Made of clear glass
  • For use with biological and pathological specimens
  • For storage of industrial laboratory chemicals and reagents