Fisherbrand™ Graduated AC Round Bottle with Phenolic PolyCone Lined Cap
Multipurpose containers for use with biological and pathological specimens.
Manufacturer:  Fisherbrand™ 278399
 View more versions of this product
Catalog No. 02911464B
Description
Description
  • Used for storing industrial laboratory chemicals and reagents
  • Designed with slight shoulder for easy pouring, handling
  • Facilitates easy mixing, handling, transfer of contents
  • Graduated in both milliliters and ounces
Specifications
Specifications
60mL
Phenolic
Storing dry materials
48/Cs.
Yes
38-400
2 oz.
Polycone
Glass
Graduated bottle
Black
Screw