Please login to your online account to display your discounted pricing

Whatman™ Grade 995 General Purpose Filter Paper - Circle

Educational grade filter paper recommended for general filtrations

$93.88 - $93.88

Specifications

Format Circle
Grade 995
Material Cellulose
Flow Rate Medium
For Use With (Application) General purpose and non-critical laboratory filtration procedures
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    

09-924-333

 
whatman™
WHT10318622*
320mm Pack of 100 Each for $93.88
Description & Specifications

Specifications

Format Circle
Grade 995
Material Cellulose
Flow Rate Medium
For Use With (Application) General purpose and non-critical laboratory filtration procedures

  • Smooth-surfaced wood pulp
  • High wet-strength
  • Medium flow strength