missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Whatman™ Grade 91 Wet Strengthened Qualitative Filter Papers - Sheets
High wet strength due to the addition of chemically stable resin
Supplier: Cytiva 1091930
Description
- Wet strengthened grade
- General purpose creped filter
- For less critical routine analysis
- Used to assay sucrose in cane sugar
Specifications
Grade 91 Qualitative Filter Paper Wet-Strengthened, sheet, 58 x 58 cm | |
Cellulose | |
70 sec. | |
Grade 91 | |
205 μm | |
580 mm |
Sheet | |
10 μm | |
A general purpose creped filter for less critical routine analysis. Widely used to assay sucrose in cane sugar and within pharmaceutical laboratories for routine filtration. | |
500 Pieces | |
58 x 58 cm | |
580 mm |