Please login to your online account to display your discounted pricing

Whatman™ Grade 598 1/2 Qualitative Folded Paper Filter - Circle

Decreased overall filtration time and increased total loading capacity

$123.71 - $629.84

Specifications

Format Folded
Grade 598.5, Folded (Prepleated)
Flow Rate Medium Fast, thick
Quantity 50/Pk.
Thickness 320μm
View More Specs
Products
Catalog Number Mfr. No. Diameter (Metric) Price Quantity    

09-924-196

 
whatman™
WHT10312244
125mm Pack of 50 Each for $123.71

09-924-197

 
whatman™
WHT10312247
185mm Pack of 50 Each for $151.56

09-924-198

 
whatman™
WHT10312251
240mm Pack of 50 Each for $205.22

09-924-199

 
whatman™
WHT10312256
500mm Pack of 50 Each for $629.84
Description & Specifications

Specifications

Format Folded
Grade 598.5, Folded (Prepleated)
Flow Rate Medium Fast, thick
Quantity 50/Pk.
Thickness 320μm

  • Thick filter paper
  • High loading capacity
  • Medium retention
  • Medium-fast to quick filtration speed