Please login to your online account to display your discounted pricing

Whatman™ Grade 597 Qualitative Folded Paper Filter- Sheet

For many routine analytical applications in different industries

$566.50 - $3,458.64

Specifications

Format Folded
Grade 597
Flow Rate Medium Fast
Length (Metric) 580mm
Particle Retention Medium Fine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

09-924-193

 
whatman™
WHT10311887
500 pieces Pack of 500 Each for $3,458.64

09-924-194

 
whatman™
WHT10311897
100 pieces Pack of 100 Each for $566.50
Description & Specifications

Specifications

Format Folded
Grade 597
Flow Rate Medium Fast
Length (Metric) 580mm
Particle Retention Medium Fine
Width (Metric) 580mm

  • Medium fast filter paper
  • Medium to fine particle retention
  • Filters fat content for food testing
  • Removes CO2, turbidity in beverage analysis