Please login to your online account to display your discounted pricing

GE Healthcare, Whatman Grade 1573-1/2 Qualitative Filter Papers

A medium fast filter paper with high wet strength. This paper has the same chemical resistance characteristics as Grade 1573

$126.80 - $126.80

Specifications

Format Circle
Grade 1573-1/2
Material Cellulose
Flow Rate Fast
Pore Size (Metric) 12 to 25μm
View More Specs
Products
Catalog Number Mfr. No. Diameter (English) Diameter (Metric) Price Quantity    

0994729

 
ge healthcare, whatman
10314745
5.9 in. 150mm Pack of 100 Each for $126.80
Description & Specifications

Specifications

Format Circle
Grade 1573-1/2
Material Cellulose
Flow Rate Fast
Pore Size (Metric) 12 to 25μm
Thickness 170μm

  • Savings in time required to quadrant-fold circles to fit conical filter funnels in repetitive or multiple analyses.
  • Decreased overall filtration time because of the extra surface area exposed; the normal slow down of filtration speed due to the loading of particulate is postponed.
  • Increased total loading capacity as more filter area is available.
  • Maintained flow rate due to the reduction in filter paper contact with funnel side and the self-supporting shape of the filter itself.
  • The prepleating does not significantly affect any of the technical data, and the same figures may be used for the flat circles.