BUCHI™ FlashPure Silica Flash Cartridges
High resolution and outstanding loading capacity
Manufacturer:  BUCHI™ 145155972
 View more versions of this product
Catalog No. 05-404-820
Includes: 3 Pieces
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
1500g
3 pieces
75psig
For flash chromatography using ISCO and Biotage instruments
Silica - GraceResolv™
Reveleris™
Documents