BUCHI FlashPure Flash Chromatography Cartridges
FlashPure flash chromatography cartridges offer high resolution and outstanding loading capacity.
Supplier: BUCHI 11067709
Includes: 6 Pieces
Description
- High resolution of separation due to high-quality silica
- Increased fraction purity due to a better separation
- Faster evaporation of smaller fraction volumes
- Reproducible performance from batch to batch
- Increased sample throughput due to high sample loading capacity
- Solvent saving: required purification yield is achieved faster
Specifications
Silica - Normal Phase | |
6 Pc. | |
For Flash Chromatography Using ISCO and Biotage Instruments |
FlashPure | |
220 g | |
75 psig |