Please login to your online account to display your discounted pricing

Vydac™ 218 MS Polymeric C18 Analytical HPLC Columns

Suitable for small peptides to large intact, undenatured proteins

Manufacturer: grace davison disc sci  5107955

 View all options for this product

Catalog No. 07-650-110


Description & Specifications


Column Type Vydac™ 218 MS C18
For Use With LC/MS separation
Type HPLC Column
Carbon Load 8%
USP Type L1
Endcapped Yes
Packing Material Vydac™ 218 MS C18
Particle Shape Spherical
Particle Size 5μm
Pore Size 300Å
Stationary Phase Vydac™ 218 MS C18
Surface Area 60 to 110m2/g
Film Thickness 5μm
Length (Metric) 50mm
Support Material Silica
Technique Reverse phase HPLC
Diameter (Metric) Inner 4.6mm
Phase Polymeric

  • Unique selectivity reveals peaks otherwise masked by other C18 columns
  • Excellent peak shape with little or no TFA
  • High protein recoveries make scale-up easy