missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

GR 89696 fumarate, Tocris

Subtype-selective κ opioid agonist

$205.70 - $862.95

Chemical Identifiers

Assay Percent Range >99%
Molecular Formula C19H25Cl2N3O3·C4H4O4
Formula Weight 530.4
Synonym 4-[(3, 4-Dichlorophenyl)acetyl]-3-(1-pyrrolidinylmethyl)-1-piperazinecarboxylic acid methyl ester fumarate
Chemical Name or Material GR 89696 fumarate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
148350 R & D Systems
148350
50mg Each for $862.95
148310 R & D Systems
148310
10mg Each for $205.70
Specifications Description & Specifications

Chemical Identifiers

>99%
530.4
GR 89696 fumarate
C19H25Cl2N3O3·C4H4O4
4-[(3, 4-Dichlorophenyl)acetyl]-3-(1-pyrrolidinylmethyl)-1-piperazinecarboxylic acid methyl ester fumarate

Specifications

50mg
>99%
530.4
GR 89696 fumarate
126766-32-3
C19H25Cl2N3O3·C4H4O4
4-[(3, 4-Dichlorophenyl)acetyl]-3-(1-pyrrolidinylmethyl)-1-piperazinecarboxylic acid methyl ester fumarate

Safety and Handling

Recommended Storage : Store at Room Temperature