Please login to your online account to display your discounted pricing

GR 55562 dihydrochloride, Tocris


5-HT1B antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS105410

 View more versions of this product

Catalog No. 105410

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material GR 55562 dihydrochloride
Quantity 10mg
CAS 159533-25-2
Recommended Storage Desiccate at +4°C
Assay Percent Range >98%
Molecular Formula C23H25N3O2·2HCl
Formula Weight 448.39
Synonym 3-[3-(Dimethylamino)propyl]-4-hydroxy-N-[4-(4-pyridinyl)phenyl]benzamide dihydrochloride

Specifications

Chemical Name or Material GR 55562 dihydrochloride
Assay Percent Range >98%
Molecular Formula C23H25N3O2·2HCl
Formula Weight 448.39
Synonym 3-[3-(Dimethylamino)propyl]-4-hydroxy-N-[4-(4-pyridinyl)phenyl]benzamide dihydrochloride
Accessories

Desiccate at +4°C