Please login to your online account to display your discounted pricing

GR 46611, Tocris


5-HT1D agonist

Quantity 50mg

Manufacturer: r & d systems  086450

 View more versions of this product

Catalog No. 086450

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 377.49
Chemical Name or Material GR 46611
Molecular Formula C23H27N3O2
CAS 185259-85-2
Synonym 3-[3-(2-Dimethylaminoethyl)-1H-indol-5-yl]-N-(4-methoxybenzyl)acrylamide
Assay Percent Range >98%
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material GR 46611
Assay Percent Range >98%
Molecular Formula C23H27N3O2
Formula Weight 377.49
Synonym 3-[3-(2-Dimethylaminoethyl)-1H-indol-5-yl]-N-(4-methoxybenzyl)acrylamide
Accessories

Store at Room Temperature