Please login to your online account to display your discounted pricing

GR 46611, Tocris


5-HT1D agonist

Quantity 10mg

Manufacturer: r & d systems  086410

 View more versions of this product

Catalog No. 086410

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 377.49
Assay Percent Range >98%
Molecular Formula C23H27N3O2
CAS 185259-85-2
Synonym 3-[3-(2-Dimethylaminoethyl)-1H-indol-5-yl]-N-(4-methoxybenzyl)acrylamide
Recommended Storage Store at Room Temperature
Chemical Name or Material GR 46611

Specifications

Chemical Name or Material GR 46611
Assay Percent Range >98%
Molecular Formula C23H27N3O2
Formula Weight 377.49
Synonym 3-[3-(2-Dimethylaminoethyl)-1H-indol-5-yl]-N-(4-methoxybenzyl)acrylamide
Accessories

Store at Room Temperature