Please login to your online account to display your discounted pricing

GR 46611, Tocris

5-HT1D agonist

$225.77 - $924.23

Chemical Identifiers

Chemical Name or Material GR 46611
Assay Percent Range >98%
Molecular Formula C23H27N3O2
Formula Weight 377.49
Synonym 3-[3-(2-Dimethylaminoethyl)-1H-indol-5-yl]-N-(4-methoxybenzyl)acrylamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

086450

 
r & d systems
086450
50mg Each for $924.23

086410

 
r & d systems
086410
10mg Each for $225.77
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 377.49
Chemical Name or Material GR 46611
Molecular Formula C23H27N3O2
CAS 185259-85-2
Synonym 3-[3-(2-Dimethylaminoethyl)-1H-indol-5-yl]-N-(4-methoxybenzyl)acrylamide
Assay Percent Range >98%
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature