Please login to your online account to display your discounted pricing

GR 159897, Tocris


Non-peptide, potent NK2 antagonist

Quantity 10mg

Manufacturer: r & d systems  RDS127410

 View more versions of this product

Catalog No. 127410

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 414.54
Quantity 10mg
Chemical Name or Material GR 159897
Assay Percent Range >97%
CAS 158848-32-9
Molecular Formula C23H27FN2O2S
Synonym 5-Fluoro-3-[2-[4-methoxy-4-[[(R)-phenylsulphinyl]methyl]-1-piperidinyl]ethyl]-1H-indole
Recommended Storage Desiccate at -20°C

Specifications

Chemical Name or Material GR 159897
Assay Percent Range >97%
Molecular Formula C23H27FN2O2S
Formula Weight 414.54
Synonym 5-Fluoro-3-[2-[4-methoxy-4-[[(R)-phenylsulphinyl]methyl]-1-piperidinyl]ethyl]-1H-indole
Accessories

Desiccate at -20°C