Please login to your online account to display your discounted pricing

GR 144053 trihydrochloride, Tocris


Glycoprotein IIb/IIIa (integrin αIibβ3) receptor antagonist; Antithrombotic

Quantity 10mg

Manufacturer: r & d systems  126310

 View more versions of this product

Catalog No. 126310

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 454.83
Quantity 10mg
Chemical Name or Material GR 144053 trihydrochloride
Assay Percent Range >99%
Molecular Formula C18H27N5O2·3HCl
CAS 1215333-48-4
Synonym 4-[4-[4-(Aminoiminomethyl)phenyl]-1-piperazinyl]-1-piperidineacetic acid trihydrochloride
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material GR 144053 trihydrochloride
Assay Percent Range >99%
Molecular Formula C18H27N5O2·3HCl
Formula Weight 454.83
Synonym 4-[4-[4-(Aminoiminomethyl)phenyl]-1-piperazinyl]-1-piperidineacetic acid trihydrochloride
Accessories

Desiccate at +4°C