Please login to your online account to display your discounted pricing

GR 144053 trihydrochloride, Tocris

Glycoprotein IIb/IIIa (integrin αIibβ3) receptor antagonist; Antithrombotic

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material GR 144053 trihydrochloride
Assay Percent Range >99%
Molecular Formula C18H27N5O2·3HCl
Formula Weight 454.83
Synonym 4-[4-[4-(Aminoiminomethyl)phenyl]-1-piperazinyl]-1-piperidineacetic acid trihydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

126350

 
r & d systems
126350
50mg Each for $844.73

126310

 
r & d systems
126310
10mg Each for $201.36
Description & Specifications

Specifications

Formula Weight 454.83
Quantity 50mg
Molecular Formula C18H27N5O2·3HCl
CAS 1215333-48-4
Assay Percent Range >99%
Chemical Name or Material GR 144053 trihydrochloride
Synonym 4-[4-[4-(Aminoiminomethyl)phenyl]-1-piperazinyl]-1-piperidineacetic acid trihydrochloride
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C