Please login to your online account to display your discounted pricing

GR 135531, Tocris


High affinity melatonin MT3 ligand

Quantity 50mg

Manufacturer: r & d systems  089650

 View more versions of this product

Catalog No. 089650

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 275.31
Chemical Name or Material GR 135531
Assay Percent Range >97%
Molecular Formula C14H17N3O3
CAS 190277-13-5
Synonym 5-Methoxycarbonylamino-N-acetyltryptamine
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material GR 135531
Assay Percent Range >97%
Molecular Formula C14H17N3O3
Formula Weight 275.31
Synonym 5-Methoxycarbonylamino-N-acetyltryptamine
Accessories

Store at Room Temperature